Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc24
Entrez Gene ID
541495
Gene Name
coiled-coil domain containing 24
Gene Aliases
wu:fc21f03, zgc:113043
Chromosome Location
Chr.2: 33422353 - 33455277 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326378.1 NP_001313307.1 1 - 2 214 110
XM_005163498.4 XP_005163555.1 2 - 3 272 110
GenBank mRNA BC091921.1 - 1 - 2 441 110

View Full Product Details