Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc24
Entrez Gene ID
541495
Gene Name
coiled-coil domain containing 24
Gene Aliases
wu:fc21f03, zgc:113043
Chromosome Location
Chr.2: 33422353 - 33455277 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326378.1 NP_001313307.1 4 - 5 633 96
XM_005163498.4 XP_005163555.1 5 - 6 691 96
XM_009298618.3 XP_009296893.1 3 - 4 621 96
GenBank mRNA BC091921.1 - 4 - 5 860 96
DT063873.1 - 4 - 5 463 96

View Full Product Details