Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm39b
Entrez Gene ID
541556
Gene Name
RNA binding motif protein 39b
Gene Aliases
rnpc2l, wu:fa97g07, wu:fb09c08, zgc:112139, zgc:113117
Chromosome Location
Chr.8: 37231752 - 37249832 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014370.1 NP_001014392.1 3 - 4 358 73
XM_005167222.4 XP_005167279.1 2 - 3 308 73
XM_021478251.1 XP_021333926.1 2 - 3 308 73
GenBank mRNA BC091794.1 - 3 - 4 358 73
BC093217.1 - 2 - 3 289 73

View Full Product Details