Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pecr
Entrez Gene ID
550422
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Gene Aliases
zgc:112155
Chromosome Location
Chr.11: 30355441 - 30364847 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017727.1 NP_001017727.1 4 - 5 544 95
XM_005169364.4 XP_005169421.1 4 - 5 514 95
XM_005169365.4 XP_005169422.1 4 - 5 551 95
GenBank mRNA BC093230.1 - 4 - 5 544 95

View Full Product Details