Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acbd5a
Entrez Gene ID
553674
Gene Name
acyl-CoA binding domain containing 5a
Gene Aliases
zgc:112043
Chromosome Location
Chr.24: 5915892 - 5933146 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020647.1 NP_001018483.1 9 - 10 1195 110
XM_005162509.4 XP_005162566.1 9 - 10 1217 110
XM_009297238.3 XP_009295513.1 10 - 11 1250 110
XM_009297239.3 XP_009295514.1 10 - 11 1235 110
XM_009297241.3 XP_009295516.1 10 - 11 1292 110
XM_021470173.1 XP_021325848.1 10 - 11 1392 110
XM_021470174.1 XP_021325849.1 10 - 11 1247 110
GenBank mRNA BC095655.1 - 9 - 10 1195 110

View Full Product Details