Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pak2b
Entrez Gene ID
325034
Gene Name
p21 protein (Cdc42/Rac)-activated kinase 2b
Gene Aliases
CH73-159L23.1, wu:fc52b03, zgc:113857
Chromosome Location
Chr.22: 2897166 - 2929156 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025456.1 NP_001020627.1 2 - 3 491 115
XM_005170360.4 XP_005170417.1 2 - 3 368 115
GenBank mRNA BC081414.1 - 2 - 3 348 115
BC095627.1 - 2 - 3 462 115
BC096967.1 - 2 - 3 491 115

View Full Product Details