Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgsn
Entrez Gene ID
567024
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
gluld1, lgs, zgc:136604
Chromosome Location
Chr.11: 11263451 - 11266878 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031674.2 NP_001026844.2 3 - 4 978 72
GenBank mRNA BC114224.1 - 3 - 4 978 72
DQ122929.1 - 3 - 4 982 72

View Full Product Details