Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
smyd1b
Entrez Gene ID
569027
Gene Name
SET and MYND domain containing 1b
Gene Aliases
sth, symd1
Chromosome Location
Chr.8: 1015243 - 1051544 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039636.1 NP_001034725.1 3 - 4 517 55
XM_021478292.1 XP_021333967.1 3 - 4 623 55
GenBank mRNA BC115351.1 - 3 - 4 521 55
BC163095.1 - 3 - 4 606 55
DQ323979.1 - 3 - 4 517 55
DQ323980.1 - 3 - 4 517 55

View Full Product Details