The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044960.2 | NP_001038425.2 | 25 - 26 | 2214 | 86 |
XM_005164746.4 | XP_005164803.1 | 25 - 26 | 2390 | 86 | |
XM_009300285.3 | XP_009298560.1 | 26 - 27 | 2416 | 86 | |
XM_009300286.3 | XP_009298561.1 | 26 - 27 | 2413 | 86 | |
XM_009300287.3 | XP_009298562.1 | 26 - 27 | 2414 | 86 | |
XM_009300288.3 | XP_009298563.1 | 25 - 26 | 2392 | 86 |