Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stc1
Entrez Gene ID
751747
Gene Name
stanniocalcin 1
Gene Aliases
zgc:153307
Chromosome Location
Chr.8: 50322782 - 50346235 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045457.1 NP_001038922.1 1 - 2 271 65
GenBank mRNA BC122228.1 - 1 - 2 271 65
BC165445.1 - 1 - 2 148 65

View Full Product Details