Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col27a1b
Entrez Gene ID
560145
Gene Name
collagen, type XXVII, alpha 1b
Gene Aliases
col27a1
Chromosome Location
Chr.5: 93570 - 181338 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080575.1 NP_001074044.1 27 - 28 2538 111
XM_017356118.2 XP_017211607.1 27 - 28 2956 111
XM_021476201.1 XP_021331876.1 7 - 8 318 111
GenBank mRNA EF032484.1 - 27 - 28 2538 111

View Full Product Details