Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pcsk5b
Entrez Gene ID
559556
Gene Name
proprotein convertase subtilisin/kexin type 5b
Gene Aliases
pcsk5, zgc:165659
Chromosome Location
Chr.8: 38649986 - 38842320 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083829.1 NP_001077298.1 7 - 8 1301 81
GenBank mRNA BC162145.1 - 7 - 8 1159 81
DQ983233.1 - 7 - 8 1301 81

View Full Product Details