Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nitr14b
Entrez Gene ID
100148851
Gene Name
novel immune-type receptor 14b
Gene Aliases
si:dkey-149k8.5
Chromosome Location
Chr.14: 27313 - 30107 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083870.1 NP_001077339.1 2 - 3 415 143
GenBank mRNA CU458983.1 - 2 - 3 415 143

View Full Product Details