Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
elavl3
Entrez Gene ID
30732
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HuC, elrc, id:ibd1248, wu:fb77b03, zHuC
Chromosome Location
Chr.3: 46798511 - 46817871 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131449.1 NP_571524.1 5 - 6 739 86
XM_017353191.2 XP_017208680.1 5 - 6 1115 86
GenBank mRNA U62018.1 - 5 - 6 739 86

View Full Product Details