Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
efnb1
Entrez Gene ID
114377
Gene Name
ephrin-B1
Gene Aliases
-
Chromosome Location
Chr.5: 16733 - 74405 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131805.1 NP_571880.1 4 - 5 641 69
GenBank mRNA AF375224.1 - 4 - 5 641 69
BC163323.1 - 4 - 5 737 69
BC163326.1 - 4 - 5 737 69
CU638799.1 - 4 - 5 1537 69

View Full Product Details