Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpib
Entrez Gene ID
246095
Gene Name
glucose-6-phosphate isomerase b
Gene Aliases
PGI, gpi, pgi-2
Chromosome Location
Chr.18: 402038 - 423921 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ306396.1 - 10 - 11 968 93

View Full Product Details