Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gria3a
Entrez Gene ID
170452
Gene Name
glutamate receptor, ionotropic, AMPA 3a
Gene Aliases
gria3.1, zfGRIA3[a]
Chromosome Location
Chr.5: 23352348 - 23429545 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198339.1 NP_938153.1 14 - 15 2471 69
XM_005165096.4 XP_005165153.1 14 - 15 2931 69
GenBank mRNA AF525745.1 - 14 - 15 2471 69

View Full Product Details