Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glud1b
Entrez Gene ID
373092
Gene Name
glutamate dehydrogenase 1b
Gene Aliases
cb719, glud1, wu:fb16e02, wu:fb58f12, wu:fe37f03, wu:fj43f02, zgc:192851, zgc:55630
Chromosome Location
Chr.12: 25845494 - 25881944 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199545.4 NP_955839.2 6 - 7 1088 83
GenBank mRNA AY577003.1 - 6 - 7 1088 83
BC063940.1 - 6 - 7 952 83

View Full Product Details