Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
upp2
Entrez Gene ID
393113
Gene Name
uridine phosphorylase 2
Gene Aliases
uppl, zgc:55883
Chromosome Location
Chr.9: 16097812 - 16108999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200144.1 NP_956438.1 3 - 4 505 127
GenBank mRNA BC044525.1 - 3 - 4 505 127

View Full Product Details