Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trim47
Entrez Gene ID
393254
Gene Name
tripartite motif containing 47
Gene Aliases
fc04b02, wu:fc04b02, zgc:56368
Chromosome Location
Chr.10: 26118142 - 26127058 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200284.1 NP_956578.1 2 - 3 451 111
GenBank mRNA BC049412.1 - 2 - 3 451 111

View Full Product Details