Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc2a12
Entrez Gene ID
393510
Gene Name
solute carrier family 2 member 12
Gene Aliases
zgc:66027, zgc:85776
Chromosome Location
Chr.23: 31608255 - 31633625 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200538.1 NP_956832.1 2 - 3 160 150
XM_005162304.4 XP_005162361.1 3 - 4 354 150
GenBank mRNA BC056306.1 - 2 - 3 160 150
BC067616.1 - 2 - 3 168 150

View Full Product Details