Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spag7
Entrez Gene ID
394100
Gene Name
sperm associated antigen 7
Gene Aliases
zgc:66397
Chromosome Location
Chr.5: 68059924 - 68074592 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201125.1 NP_957419.1 4 - 5 346 115
GenBank mRNA BC054697.1 - 4 - 5 346 115

View Full Product Details