Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bnip2
Entrez Gene ID
394197
Gene Name
BCL2 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.25: 34246448 - 34270735 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201218.1 NP_957512.1 3 - 4 347 62
XM_009298160.3 XP_009296435.1 4 - 5 1287 62
XM_009298162.3 XP_009296437.1 4 - 5 1284 62
GenBank mRNA AY423003.1 - 3 - 4 347 62
BC114255.1 - 4 - 5 392 62
BC124462.1 - 4 - 5 638 62
BC134072.1 - 4 - 5 1121 62

View Full Product Details