Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
mttp
Entrez Gene ID
406207
Gene Name
microsomal triglyceride transfer protein
Gene Aliases
mtp, stl, wu:fd36b01, zgc:111876, zgc:136353
Chromosome Location
Chr.1: 12107680 - 12126541 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212970.2 NP_998135.2 13 - 14 2037 61
GenBank mRNA AJ428850.1 - 13 - 14 1930 61

View Full Product Details