Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vps26bl
Entrez Gene ID
436688
Gene Name
vacuolar protein sorting 26 homolog B, like
Gene Aliases
zgc:92676
Chromosome Location
Chr.10: 31800212 - 31805921 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002415.2 NP_001002415.1 5 - 6 1094 93
GenBank mRNA BC076160.1 - 5 - 6 1096 93

View Full Product Details