Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ralba
Entrez Gene ID
445255
Gene Name
v-ral simian leukemia viral oncogene homolog Ba (ras related)
Gene Aliases
ralb, zgc:100801
Chromosome Location
Chr.9: 36895188 - 36924389 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003649.1 NP_001003649.1 2 - 3 309 72
XM_021478640.1 XP_021334315.1 2 - 3 336 72
GenBank mRNA BC078184.1 - 2 - 3 309 72

View Full Product Details