Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
azin1b
Entrez Gene ID
317764
Gene Name
antizyme inhibitor 1b
Gene Aliases
azin1, oazin, wu:fj32d12
Chromosome Location
Chr.19: 67126 - 80685 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007159.2 NP_001007160.2 11 - 12 1596 93
GenBank mRNA BC061960.1 - 9 - 10 1252 93

View Full Product Details