Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pih1d2
Entrez Gene ID
494086
Gene Name
PIH1 domain containing 2
Gene Aliases
zgc:101725
Chromosome Location
Chr.15: 18138518 - 18162650 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008629.1 NP_001008629.1 1 - 2 424 72
XM_009291562.1 XP_009289837.1 1 - 2 422 72
XM_021481399.1 XP_021337074.1 1 - 2 419 72
XM_021481401.1 XP_021337076.1 1 - 2 419 72
XM_021481402.1 XP_021337077.1 1 - 2 419 72
GenBank mRNA BC086728.1 - 1 - 2 424 72

View Full Product Details