Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cuedc2
Entrez Gene ID
334351
Gene Name
CUE domain containing 2
Gene Aliases
wu:fi42e02, zgc:92255
Chromosome Location
Chr.13: 29925402 - 29936259 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017994.2 NP_001017994.1 5 - 6 706 130
XM_005156782.2 XP_005156839.1 6 - 7 778 130
GenBank mRNA AY394932.1 - 6 - 7 787 130
BC081633.1 - 5 - 6 706 130

View Full Product Details