Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nitr11a
Entrez Gene ID
558243
Gene Name
novel immune-type receptor 11a
Gene Aliases
nitr11a_1
Chromosome Location
Chr.7: 71846 - 73750 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025501.1 NP_001020672.1 2 - 3 383 92

View Full Product Details