Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kera
Entrez Gene ID
567017
Gene Name
keratocan
Gene Aliases
si:dkeyp-38g8.3, zgc:136259
Chromosome Location
Chr.4: 16337156 - 16345059 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025548.2 NP_001020719.1 2 - 3 1003 64
XM_005164765.2 XP_005164822.1 2 - 3 981 64
GenBank mRNA BC115058.1 - 2 - 3 1003 64

View Full Product Details