Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grpel1
Entrez Gene ID
559037
Gene Name
GrpE-like 1, mitochondrial
Gene Aliases
zgc:123352
Chromosome Location
Chr.7: 39744244 - 39751529 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037384.1 NP_001032461.1 1 - 2 109 64
GenBank mRNA AI626746.1 - 1 - 2 74 64
BC108002.1 - 1 - 2 109 64

View Full Product Details