Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smtlb
Entrez Gene ID
553408
Gene Name
somatolactin beta
Gene Aliases
slb
Chromosome Location
Chr.10: 39314578 - 39321366 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037674.1 NP_001032763.1 3 - 4 347 76
GenBank mRNA AY221126.1 - 3 - 4 347 76

View Full Product Details