Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
foxp1b
Entrez Gene ID
569047
Gene Name
forkhead box P1b
Gene Aliases
foxp1, im:7139058, wu:fc83a06
Chromosome Location
Chr.6: 43598065 - 43890849 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039637.1 NP_001034726.1 5 - 6 445 59
XM_005166055.4 XP_005166112.1 6 - 7 828 59
XM_009302561.3 XP_009300836.1 6 - 7 830 59
XM_021476910.1 XP_021332585.1 6 - 7 527 59
XM_021476911.1 XP_021332586.1 5 - 6 788 59
XM_021476912.1 XP_021332587.1 3 - 4 620 59
XM_021476913.1 XP_021332588.1 2 - 3 183 59
XM_021476914.1 XP_021332589.1 2 - 3 280 59
XM_021476916.1 XP_021332591.1 2 - 3 417 59
XM_021476917.1 XP_021332592.1 6 - 7 829 59
XM_021476918.1 XP_021332593.1 1 - 2 156 59
GenBank mRNA BC163445.1 - 2 - 3 196 59
DQ333303.2 - 5 - 6 445 59

View Full Product Details