Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nfatc1
Entrez Gene ID
568315
Gene Name
nuclear factor of activated T cells 1
Gene Aliases
si:ch211-198c22.2
Chromosome Location
Chr.19: 22216778 - 22303614 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045159.1 NP_001038624.1 2 - 3 1375 71
GenBank mRNA DQ530435.1 - 2 - 3 1375 71

View Full Product Details