Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dmrt2b
Entrez Gene ID
558970
Gene Name
doublesex and mab-3 related transcription factor 2b
Gene Aliases
-
Chromosome Location
Chr.6: 202227 - 208456 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079976.1 NP_001073445.1 1 - 2 308 79
GenBank mRNA BC163825.1 - 1 - 2 394 79
BC163830.1 - 1 - 2 394 79
DQ307066.1 - 1 - 2 308 79

View Full Product Details