Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrxn2b
Entrez Gene ID
560420
Gene Name
neurexin 2b
Gene Aliases
-
Chromosome Location
Chr.7: 427503 - 624158 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079984.1 NP_001073453.1 9 - 10 4169 135
GenBank mRNA DQ641430.1 - 9 - 10 4169 135
DQ641431.1 - 2 - 3 1444 135

View Full Product Details