Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrxn1a
Entrez Gene ID
565531
Gene Name
neurexin 1a
Gene Aliases
-
Chromosome Location
Chr.12: 114926 - 403053 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080021.2 NP_001073490.1 19 - 20 4667 65
NM_001346923.1 NP_001333852.1 3 - 4 1559 65
XM_005156209.4 XP_005156266.1 19 - 20 4661 65
XM_009306641.3 XP_009304916.1 20 - 21 4688 65
XM_009306642.3 XP_009304917.1 20 - 21 4685 65
XM_009306643.3 XP_009304918.1 20 - 21 4687 65
XM_009306644.3 XP_009304919.1 20 - 21 4676 65
XM_009306645.3 XP_009304920.1 19 - 20 4676 65
XM_009306646.3 XP_009304921.1 19 - 20 4673 65
XM_009306648.3 XP_009304923.1 19 - 20 4670 65
XM_009306649.3 XP_009304924.1 19 - 20 4664 65
XM_009306650.3 XP_009304925.1 19 - 20 4661 65
XM_009306651.3 XP_009304926.1 19 - 20 4648 65
XM_021480136.1 XP_021335811.1 18 - 19 4658 65
XM_021480137.1 XP_021335812.1 19 - 20 4648 65
XM_021480138.1 XP_021335813.1 17 - 18 4628 65
XM_021480139.1 XP_021335814.1 18 - 19 4622 65
XM_021480140.1 XP_021335815.1 17 - 18 4612 65
XM_021480143.1 XP_021335818.1 17 - 18 4618 65
XM_021480144.1 XP_021335819.1 15 - 16 4561 65
GenBank mRNA AB214179.1 - 4 - 5 1067 65
DQ641424.1 - 18 - 19 4616 65
DQ641425.1 - 3 - 4 1559 65

View Full Product Details