Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kdm6al
Entrez Gene ID
569207
Gene Name
lysine (K)-specific demethylase 6A, like
Gene Aliases
utxl1, uty, zgc:158313
Chromosome Location
Chr.6: 59851523 - 59895080 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080193.1 NP_001073662.1 10 - 11 1125 57
XM_005168796.4 XP_005168853.1 10 - 11 3826 57
GenBank mRNA BC066564.1 - 10 - 11 1444 57
BC129198.1 - 10 - 11 1125 57

View Full Product Details