Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sema4e
Entrez Gene ID
796377
Gene Name
semaphorin 4e
Gene Aliases
sema7, semaZ7
Chromosome Location
Chr.2: 49568980 - 49603676 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083865.1 NP_001077334.1 11 - 12 1446 67
NM_131049.1 NP_571124.1 13 - 14 1990 67
XM_005168429.4 XP_005168486.1 13 - 14 1979 67
XM_017359054.2 XP_017214543.1 13 - 14 1958 67
XM_021481386.1 XP_021337061.1 12 - 13 1920 67
GenBank mRNA AF073289.1 - 13 - 14 1990 67
BC078280.1 - 12 - 13 1595 67
BC155168.1 - 13 - 14 1953 67
CU458994.1 - 11 - 12 1446 67

View Full Product Details