Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ephb4a
Entrez Gene ID
30688
Gene Name
eph receptor B4a
Gene Aliases
cb646, ephB4, rtk5
Chromosome Location
Chr.5: 68407447 - 68495991 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131414.1 NP_571489.1 7 - 8 1533 92
XM_021476001.1 XP_021331676.1 7 - 8 2301 92
GenBank mRNA AJ005026.2 - 7 - 8 1533 92
KM655578.1 - 7 - 8 1442 92

View Full Product Details