Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cldnc
Entrez Gene ID
81582
Gene Name
claudin c
Gene Aliases
-
Chromosome Location
Chr.21: 25754276 - 25756119 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131764.1 NP_571839.1 1 - 1 850 63
GenBank mRNA AF359432.1 - 1 - 1 850 63
BC065424.1 - 1 - 1 705 63

View Full Product Details