Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
selenot1b
Entrez Gene ID
352921
Gene Name
selenoprotein T, 1b
Gene Aliases
cb698, selt1b, sept1b, wu:fb51a03, wu:fb51b01
Chromosome Location
Chr.18: 40527 - 51890 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178292.5 NP_840077.3 1 - 2 227 77
GenBank mRNA BC053147.1 - 1 - 2 174 77
CK870132.1 - 1 - 2 194 77
CN014436.1 - 1 - 2 259 77
EH502237.1 - 1 - 2 218 77

View Full Product Details