Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
selenot1b
Entrez Gene ID
352921
Gene Name
selenoprotein T, 1b
Gene Aliases
cb698, selt1b, sept1b, wu:fb51a03, wu:fb51b01
Chromosome Location
Chr.18: 40527 - 51890 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178292.5 NP_840077.3 2 - 3 338 73
GenBank mRNA CK870132.1 - 2 - 3 305 73
EH502237.1 - 2 - 3 329 73

View Full Product Details