Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
selenot1b
Entrez Gene ID
352921
Gene Name
selenoprotein T, 1b
Gene Aliases
cb698, selt1b, sept1b, wu:fb51a03, wu:fb51b01
Chromosome Location
Chr.18: 40527 - 51890 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178292.5 NP_840077.3 5 - 6 698 64
GenBank mRNA BC053147.1 - 5 - 6 645 64
CK870132.1 - 5 - 6 665 64
EH502237.1 - 5 - 6 689 64
EH522934.1 - 1 - 2 566 64

View Full Product Details