Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cel.1
Entrez Gene ID
322481
Gene Name
carboxyl ester lipase, tandem duplicate 1
Gene Aliases
Cel, fb62e11, wu:fb62e11, zgc:66457, zgc:92042
Chromosome Location
Chr.21: 11401244 - 11411362 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199607.2 NP_955901.2 2 - 3 238 123
GenBank mRNA BC055668.1 - 2 - 3 238 123
BC079529.1 - 2 - 3 229 123

View Full Product Details