Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnaja3a
Entrez Gene ID
335894
Gene Name
DnaJ heat shock protein family (Hsp40) member A3a
Gene Aliases
wu:fj36c02
Chromosome Location
Chr.3: 59528 - 81094 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201313.1 NP_958470.1 8 - 9 1158 61
XM_005168471.4 XP_005168528.1 8 - 9 1166 61
GenBank mRNA BC047809.1 - 8 - 9 1158 61
BC066630.1 - 8 - 9 1127 61
BC165728.1 - 8 - 9 1155 61

View Full Product Details