Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eif2s3
Entrez Gene ID
327341
Gene Name
eukaryotic translation initiation factor 2, subunit 3 gamma
Gene Aliases
Su(var)3-9, fi03g03, wu:fi03g03, wu:fi37d04, zgc:63805
Chromosome Location
Chr.24: 16983426 - 17002950 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212711.3 NP_997876.2 1 - 2 374 91
GenBank mRNA AY648723.1 - 1 - 2 374 91
BC055248.1 - 1 - 2 108 91

View Full Product Details