Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glo1
Entrez Gene ID
368213
Gene Name
glyoxalase 1
Gene Aliases
cb554, wu:fb82g09, zgc:66035
Chromosome Location
Chr.13: 44788077 - 44808657 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213151.1 NP_998316.1 5 - 6 494 108
GenBank mRNA BC062383.1 - 5 - 6 494 108

View Full Product Details