Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc37a4a
Entrez Gene ID
406465
Gene Name
solute carrier family 37 member 4a
Gene Aliases
fc25d06, slc37a4, wu:fc25d06, zgc:77098
Chromosome Location
Chr.15: 9294596 - 9315555 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214738.1 NP_999903.1 6 - 7 1038 59
XM_009291487.3 XP_009289762.1 7 - 8 1265 59
XM_021481373.1 XP_021337048.1 7 - 8 1269 59
GenBank mRNA BC066719.1 - 6 - 7 1038 59

View Full Product Details